Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmga1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025427.3 | NP_001020598.1 | 5 | 716 | ||
NM_001039356.2 | NP_001034445.1 | 6 | 831 | |||
NM_001166535.1 | NP_001160007.1 | 5 | 754 | |||
NM_001166536.1 | NP_001160008.1 | 5 | 714 | |||
NM_001166537.1 | NP_001160009.1 | 5 | 1008 | |||
NM_001166539.1 | NP_001160011.1 | 6 | 798 | |||
NM_001166540.1 | NP_001160012.1 | 5 | 721 | |||
NM_001166541.1 | NP_001160013.1 | 5 | 683 | |||
NM_001166542.1 | NP_001160014.1 | 5 | 681 | |||
NM_001166543.1 | NP_001160015.1 | 5 | 751 | |||
NM_001166544.1 | NP_001160016.1 | 5 | 713 | |||
NM_001166545.1 | NP_001160017.1 | 5 | 728 | |||
NM_001166546.1 | NP_001160018.1 | 5 | 712 | |||
NM_016660.3 | NP_057869.2 | 5 | 1041 | |||
GenBank mRNA | AK010617.1 | - | 5 | 606 | ||
AK154957.1 | - | 5 | 637 | |||
AK155661.1 | - | 5 | 635 | |||
AK160036.1 | - | 5 | 605 | |||
AK169783.1 | - | 5 | 635 | |||
AK171334.1 | - | 5 | 670 | |||
CF951929.1 | - | 5 | 607 | |||
GH454882.1 | - | 6 | 793 | |||
J04179.1 | - | 5 | 709 |
Gene Symbol: Hmga1-rs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166476.1 | NP_001159948.1 | 1 | 637 | ||
NM_001166477.1 | NP_001159949.1 | 2 | 604 | |||
GenBank mRNA | BC008125.1 | - | 2 | 589 | ||
BC013455.1 | - | 1 | 647 | 4219805 | ||
BC093496.1 | - | 1 | 604 | 3601855 |