Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FKBP1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322964.1 | NP_001309893.1 | 2, 3 | 129 | ||
NM_004116.4 | NP_004107.1 | 2, 3 | 242 | |||
NM_054033.3 | NP_473374.1 | 2, 3 | 242 | |||
NR_136539.1 | 2, 3 | 129 | ||||
XM_011532699.2 | XP_011531001.1 | 2, 3 | 191 | |||
XM_017003594.1 | XP_016859083.1 | 2, 3 | 109 | |||
XM_017003595.1 | XP_016859084.1 | 1, 2 | 90 | |||
GenBank mRNA | AF322070.1 | - | 2, 3 | 134 | ||
BC002614.1 | - | 2, 3 | 185 | 3161896 | ||
BF216711.1 | - | 2, 3 | 97 | |||
BF969580.1 | - | 2, 3 | 229 | |||
BI546013.1 | - | 2, 3 | 207 | |||
BM802489.1 | - | 2, 3 | 242 | |||
BU147063.1 | - | 2, 3 | 80 | |||
BU508033.1 | - | 2, 3 | 206 | |||
D38037.1 | - | 2, 3 | 165 | |||
L37086.1 | - | 2, 3 | 159 | |||
S69800.1 | - | 2, 3 | 165 | |||
S69815.1 | - | 2, 3 | 165 |