Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015830.1 | NP_056645.1 | 11 | 3150 | ||
GenBank mRNA | AK143711.1 | - | 12 | 3894 | ||
AK148209.1 | - | 8 | 1514 | |||
AK170543.1 | - | 11 | 3214 | |||
BC058094.1 | - | 10 | 3592 | 6824892 |