Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chek2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016681.3 | NP_057890.1 | 2 | 144 | ||
GenBank mRNA | AF086905.1 | - | 1 | 105 | ||
AK044913.1 | - | 2 | 145 | |||
AK170235.1 | - | 2 | 179 | |||
BC056617.1 | - | 2 | 168 | 4977960 |