Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Auh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016709.2 | NP_057918.2 | 4 | 401 | ||
GenBank mRNA | AK003219.1 | - | 4 | 380 | ||
AK019978.1 | - | 4 | 378 | |||
AK134886.1 | - | 4 | 373 | |||
BC026525.1 | - | 4 | 339 | |||
BC049597.1 | - | 4 | 385 | 6593786 |