Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNQ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004700.3 | NP_004691.2 | 2 | 465 | ||
NM_172163.2 | NP_751895.1 | 2 | 465 | |||
GenBank mRNA | AF105202.1 | - | 2 | 465 |