Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016740.3 | NP_058020.1 | 2 | 140 | ||
GenBank mRNA | AK002490.1 | - | 2 | 142 | ||
AK003889.1 | - | 2 | 140 | |||
AK164352.1 | - | 2 | 141 | |||
BC021916.1 | - | 2 | 119 | 4913795 | ||
BC086903.1 | - | 2 | 139 | 6764162 | ||
U41341.1 | - | 1 | 74 |