Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ube2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016786.3 | NP_058066.2 | 2 | 352 | ||
GenBank mRNA | AB011081.1 | - | 2 | 290 | ||
AK018366.1 | - | 2 | 326 | |||
AK035371.1 | - | 2 | 249 | |||
AK036068.1 | - | 2 | 293 | |||
AK048194.1 | - | 2 | 343 | |||
AK139564.1 | - | 2 | 369 | |||
AK160728.1 | - | 2 | 312 | |||
AK162142.1 | - | 2 | 293 | |||
AK166647.1 | - | 2 | 261 | |||
BC002013.1 | - | 2 | 200 | 3496393 | ||
BC085311.1 | - | 2 | 278 | 30148009 | ||
BY247382.1 | - | 2 | 353 |