Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Batf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016767.2 | NP_058047.1 | 3 | 741 | ||
GenBank mRNA | AF017021.1 | - | 3 | 741 | ||
AK018587.1 | - | 3 | 782 | |||
BC132410.1 | - | 3 | 575 | 40130687 | ||
BC132412.1 | - | 3 | 575 | 40130689 |