Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNMB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278911.1 | NP_001265840.1 | 3 | 461 | ||
NM_005832.4 | NP_005823.1 | 4 | 538 | |||
NM_181361.2 | NP_852006.1 | 3 | 590 | |||
XM_011512325.2 | XP_011510627.1 | 4 | 571 | |||
GenBank mRNA | AA904191.1 | - | 4 | 376 | ||
AB193526.1 | - | 2 | 109 | |||
AB193528.1 | - | 2 | 109 | |||
AB193529.1 | - | 2 | 109 | |||
AB193530.1 | - | 2 | 109 | |||
AF099137.1 | - | 3 | 254 | |||
AF209747.1 | - | 3 | 461 | |||
AK298291.1 | - | 3 | 430 | |||
BC017825.1 | - | 3 | 452 | 4657825 | ||
BG701449.1 | - | 3 | 598 |