Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cks1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016904.1 | NP_058600.1 | 2 | 164 | ||
GenBank mRNA | AB025409.1 | - | 2 | 164 | ||
AK004101.1 | - | 2 | 150 | |||
AK010414.2 | - | 2 | 151 | |||
AK011314.1 | - | 2 | 153 | |||
AK011782.1 | - | 2 | 130 | |||
BC076579.1 | - | 2 | 156 | 6846517 |