Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164177.1 | NP_001157649.1 | 3 | 269 | ||
NM_016883.4 | NP_058579.2 | 3 | 269 | |||
GenBank mRNA | AB022022.1 | - | 3 | 225 | ||
AK009068.1 | - | 3 | 248 | |||
AK018233.1 | - | 3 | 247 | |||
AK136400.1 | - | 3 | 248 | |||
BC026931.1 | - | 3 | 236 | 3987112 | ||
BC056196.1 | - | 3 | 254 | 30019985 | ||
BY057263.1 | - | 3 | 269 | |||
CD564300.1 | - | 3 | 236 |