Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016899.4 | NP_058595.2 | 3, 4 | 629 | ||
GenBank mRNA | AB232620.1 | - | 3, 4 | 425 | ||
AF119675.1 | - | 3, 4 | 565 | |||
AK003260.1 | - | 3, 4 | 630 | |||
BC006624.1 | - | 3, 4 | 615 | 3968810 |