Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppm1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016910.3 | NP_058606.3 | 2 | 806 | ||
GenBank mRNA | AF200464.1 | - | 2 | 587 | ||
AK157818.1 | - | 2 | 794 | |||
AK166632.1 | - | 3 | 436 | |||
AK171714.1 | - | 2 | 786 | |||
BC023492.1 | - | 2 | 682 | 5026376 | ||
BC051966.1 | - | 2 | 806 | 5689560 |