Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkd2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163004.1 | NP_001156476.1 | 2 | 142 | ||
NM_016927.3 | NP_058623.2 | 2 | 142 | |||
GenBank mRNA | AF182033.1 | - | 2 | 142 | ||
AK133423.1 | - | 2 | 139 | |||
AK136119.1 | - | 246 | ||||
AK142697.1 | - | 2 | 131 | |||
AK163272.1 | - | 236 | ||||
BC138717.1 | - | 2 | 127 | 8861739 |