Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166409.2 | NP_001159881.1 | 6 | 702 | ||
NM_001166410.2 | NP_001159882.1 | 6 | 699 | |||
NM_001166411.2 | NP_001159883.1 | 6 | 699 | |||
NM_001293658.1 | NP_001280587.1 | 6 | 702 | |||
NM_016809.6 | NP_058089.2 | 6 | 702 | |||
GenBank mRNA | AB016424.1 | - | 6 | 526 | ||
AK003720.1 | - | 6 | 517 | |||
AK007354.1 | - | 6 | 517 | |||
AK049575.1 | - | 6 | 520 | |||
AK088031.1 | - | 6 | 518 | |||
AK133795.1 | - | 6 | 519 | |||
AK154044.1 | - | 6 | 521 | |||
AK167800.1 | - | 6 | 515 | |||
AK167967.1 | - | 7 | 793 | |||
AK168285.1 | - | 6 | 523 | |||
AK168338.1 | - | 7 | 793 | |||
BC006580.1 | - | 6 | 494 | 3586577 | ||
BC059098.1 | - | 6 | 508 | 4013810 | ||
BC086491.1 | - | 6 | 511 | 30434553 | ||
BC087737.1 | - | 5 | 467 | |||
BC106176.1 | - | 6 | 555 | 30146829 | ||
DV045913.1 | - | 6 | 534 |