Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy1b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161796.1 | NP_001155268.1 | 3 | 229 | ||
NM_017469.4 | NP_059497.1 | 3 | 229 | |||
GenBank mRNA | AF020339.1 | - | 3 | 203 | ||
AF297083.1 | - | 3 | 203 | |||
AK139406.1 | - | 3 | 192 | |||
AK147128.1 | - | 3 | 164 | |||
BC050945.1 | - | 3 | 207 | 6822142 | ||
BC056995.1 | - | 3 | 207 | 6822142 |