Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002250.2 | NP_002241.1 | 9 | 1701 | ||
XM_005258882.2 | XP_005258939.1 | 8 | 2487 | |||
XM_005258883.2 | XP_005258940.1 | 9 | 2579 | |||
GenBank mRNA | AF022797.1 | - | 9 | 1701 | ||
BC015337.1 | - | 9 | 1314 | 4431534 |