Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nedd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111324.2 | NP_001104794.1 | 8 | 4220 | ||
NM_017464.5 | NP_059492.3 | 7 | 4001 | |||
GenBank mRNA | AK046357.1 | - | 7 | 3967 | ||
AK083374.1 | - | 8 | 4150 | |||
AV367231.2 | - | 1 | 245 | |||
BC004696.1 | - | 3 | 2269 |