Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akap8l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017476.2 | NP_059504.2 | 10, 11 | 1385 | ||
GenBank mRNA | AB028921.1 | - | 10, 11 | 1381 | ||
AK011325.1 | - | 10, 11 | 1390 | |||
AK166526.1 | - | 11, 12 | 1301 | |||
BC003242.1 | - | 14 | ||||
BC046801.1 | - | 10, 11 | 1374 | 6403966 | ||
BC051015.1 | - | 8, 9 | 3694 |