Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Copg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017477.2 | NP_059505.1 | 24 | 3707 | ||
GenBank mRNA | AF187079.1 | - | 24 | 3706 | ||
AK077461.1 | - | 24 | 3657 | |||
AK169256.1 | - | 24 | 3753 | |||
AK172519.1 | - | 24 | 3723 | |||
AK207472.1 | - | 1 | 31 | |||
BC008553.1 | - | 16 | 2894 |