Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANXA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001156.4 | NP_001147.1 | 4 | 384 | ||
NM_001320879.1 | NP_001307808.1 | 3 | 329 | |||
NM_001320880.1 | NP_001307809.1 | 3 | 329 | |||
NM_004034.3 | NP_004025.1 | 4 | 384 | |||
XM_017016162.1 | XP_016871651.1 | 4 | 420 | |||
XM_017016163.1 | XP_016871652.1 | 4 | 420 | |||
GenBank mRNA | AB062429.1 | - | 4 | 342 | ||
AK222552.1 | - | 4 | 352 | |||
AK309141.1 | - | 3, 4 | 146 | |||
AK312447.1 | - | 4 | 357 | |||
AK313024.1 | - | 4 | 327 | |||
BC002632.2 | - | 4 | 344 | 3610541 | ||
BT007187.1 | - | 3 | 309 | |||
BX361177.2 | - | 3 | 295 | |||
CR407686.1 | - | 3 | 309 | |||
DA131184.1 | - | 3 | 302 | |||
DC367313.1 | - | 4 | 383 | |||
HY002854.1 | - | 4 | 385 | |||
J04543.1 | - | 4 | 369 |