Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018765.3 | NP_061235.2 | 3 | 441 | ||
GenBank mRNA | AF071184.1 | - | 3 | 293 | ||
AK019160.1 | - | 3 | 175 | |||
AK135215.1 | - | 3 | 441 | |||
AK159522.1 | - | 3 | 693 | |||
AK160559.1 | - | 3 | 178 | |||
AK161496.1 | - | 3 | 135 | |||
AK162009.1 | - | 3 | 183 | |||
BC034851.1 | - | 3 | 136 | 1399984 |