Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcnx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018814.3 | NP_061284.2 | 11 | 3203 | ||
GenBank mRNA | AF096286.1 | - | 5 | 1033 | ||
AK129221.1 | - | 6 | 1083 | |||
AK135724.1 | - | 5 | 659 | |||
BC057333.1 | - | 4 | 946 | |||
BC066849.1 | - | 6 | 876 | |||
BC098370.1 | - | 6 | 793 |