Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Akr1e1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018859.2 | NP_061347.2 | 2 | 184 | ||
GenBank mRNA | AI893997.1 | - | 2 | 184 | ||
AK002507.1 | - | 2 | 159 | |||
AK007924.1 | - | 2 | 161 | |||
BC012692.1 | - | 2 | 200 | 3986863 | ||
U68535.1 | - | 2 | 170 |