Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195130.1 | NP_001182059.1 | 7 | 1062 | ||
NM_018774.4 | NP_061244.1 | 7 | 1055 | |||
GenBank mRNA | AK083804.1 | - | 2 | 1202 | ||
BC057571.1 | - | 7 | 998 | 6410302 | ||
U81491.1 | - | 6 | 952 |