Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbc1d8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018775.4 | NP_061245.3 | 6, 7 | 1224 | ||
GenBank mRNA | AK138630.1 | - | 6, 7 | 1222 | ||
AK141520.1 | - | 6, 7 | 1213 | |||
AK141816.1 | - | 6, 7 | 1178 | |||
BC005421.1 | - | 3, 4 | 559 | |||
BC065081.1 | - | 6, 7 | 1158 | 6832495 | ||
U88873.1 | - | 2, 3 | 444 |