Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNQ4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004700.3 | NP_004691.2 | 10 | 1524 | ||
NM_172163.2 | NP_751895.1 | 9 | 1362 | |||
XM_017002792.1 | XP_016858281.1 | 7 | 765 | |||
GenBank mRNA | AF105202.1 | - | 10 | 1524 | ||
AK074957.1 | - | 7 | 761 |