Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpsf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018813.3 | NP_061283.2 | 3 | 351 | ||
GenBank mRNA | AF203969.1 | - | 3 | 204 | ||
AK149953.1 | - | 3 | 278 | |||
AK157205.1 | - | 3 | 293 | |||
AK158036.1 | - | 3 | 263 | |||
AK170842.1 | - | 3 | 236 | |||
AK171838.1 | - | 3 | 282 | |||
BC023297.1 | - | 3 | 237 | 4192224 | ||
BY094575.1 | - | 3 | 355 |