Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbp4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018765.3 | NP_061235.2 | 6 | 810 | ||
GenBank mRNA | AF071184.1 | - | 6 | 662 | ||
AK135215.1 | - | 6 | 810 | |||
AK159522.1 | - | 6 | 1062 | |||
AK160559.1 | - | 6 | 547 | |||
AK161496.1 | - | 6 | 504 | |||
AK162009.1 | - | 6 | 552 | |||
AK164984.1 | - | 1 | 1537 | |||
BC034851.1 | - | 6 | 505 | 1399984 | ||
U40746.1 | - | 2 | 90 |