Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Skap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018773.2 | NP_061243.1 | 11, 12 | 1140 | ||
GenBank mRNA | AB014485.1 | - | 11, 12 | 1057 | ||
AF051324.1 | - | 11, 12 | 1055 | |||
AK076000.1 | - | 11, 12 | 1085 | |||
AK144289.1 | - | 11, 12 | 1077 | |||
AK204408.1 | - | 3, 4 | 276 | |||
BC003711.1 | - | 11, 12 | 1140 | 3599914 |