Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asah2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018830.1 | NP_061300.1 | 9 | 1097 | ||
GenBank mRNA | AB037111.1 | - | 10 | 1382 | ||
AB037181.1 | - | 9 | 1097 | |||
AK034493.1 | - | 9 | 1328 | |||
AK041937.1 | - | 9 | 1286 | |||
AK046540.1 | - | 9 | 1291 | |||
AK047692.1 | - | 9 | 1285 | |||
AK080951.1 | - | 9 | 1274 | |||
AK166100.1 | - | 9 | 1345 | |||
BC137898.1 | - | 9 | 1080 | 8860917 | ||
BC145016.1 | - | 9 | 1083 | 9053564 |