Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samhd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001139520.1 | NP_001132992.1 | 3 | 434 | ||
NM_018851.3 | NP_061339.3 | 3 | 434 | |||
GenBank mRNA | AK054490.1 | - | 3 | 333 | ||
AK078936.1 | - | 3 | 377 | |||
AK149865.1 | - | 3 | 309 | |||
AK150964.1 | - | 3 | 149 | |||
AK151335.1 | - | 3 | 308 | |||
AK152815.1 | - | 3 | 149 | |||
AK153390.1 | - | 3 | 308 | |||
AK170688.1 | - | 3 | 333 | |||
AK171565.1 | - | 3 | 333 | |||
BC012721.1 | - | 3 | 371 | 4037046 | ||
BC067198.1 | - | 3 | 357 | 6826682 | ||
U15635.1 | - | 3 | 358 |