Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tyro3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290800.1 | NP_001277729.1 | 3 | 1087 | ||
NM_019392.2 | NP_062265.2 | 3 | 623 | |||
GenBank mRNA | AK141198.1 | - | 3 | 394 | ||
BC066058.1 | - | 3 | 623 | 6834015 | ||
BC082325.1 | - | 3 | 571 | 30356080 | ||
X78103.1 | - | 3 | 567 |