Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rabep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291142.1 | NP_001278071.1 | 2 | 363 | ||
NM_001291143.1 | NP_001278072.1 | 3 | 434 | |||
NM_019400.3 | NP_062273.2 | 2 | 363 | |||
GenBank mRNA | AB001750.1 | - | 3 | 270 | ||
AF248489.1 | - | 2 | 149 | |||
AK019413.1 | - | 2 | 337 | |||
AK134587.1 | - | 2 | 362 | |||
AK137236.1 | - | 2 | 328 | |||
AK151901.1 | - | 2 | 332 | |||
BC009814.1 | - | 2 | 359 | |||
BC060166.1 | - | 2 | 342 | |||
BC080274.1 | - | 2 | 336 | |||
BC129853.1 | - | 2 | 182 | 40109688 | ||
BC129854.1 | - | 2 | 182 | 40109689 | ||
BY133091.1 | - | 2 | 363 | |||
BY718394.1 | - | 2 | 362 | |||
D86066.1 | - | 2 | 199 |