Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019466.4 | NP_062339.2 | 1, 2 | 190 | ||
GenBank mRNA | AF237790.1 | - | 1, 2 | 94 | ||
AF263239.1 | - | 1, 2 | 124 | |||
AK077283.1 | - | 1, 2 | 190 | |||
BC013551.1 | - | 1, 2 | 170 | 4236038 |