Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fut2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271993.1 | NP_001258922.1 | 3 | 443 | ||
NM_018876.4 | NP_061364.2 | 2 | 339 | |||
GenBank mRNA | AF064792.1 | - | 1 | 140 | ||
AK033520.1 | - | 2 | 254 | |||
AK033586.1 | - | 2 | 341 | |||
BC138534.1 | - | 2 | 193 | 8861554 | ||
BC145831.1 | - | 2 | 193 | 40131134 |