Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pabpn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019402.2 | NP_062275.1 | 2 | 405 | ||
GenBank mRNA | AK032172.1 | - | 2 | 401 | ||
AK157143.1 | - | 2 | 394 | |||
BC049869.1 | - | 2 | 390 | |||
BC055866.1 | - | 2 | 394 | 4218469 |