Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNMB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278911.1 | NP_001265840.1 | 4 | 705 | ||
NM_005832.4 | NP_005823.1 | 5 | 782 | |||
NM_181361.2 | NP_852006.1 | 4 | 834 | |||
XM_011512325.2 | XP_011510627.1 | 5 | 815 | |||
GenBank mRNA | AB193524.1 | - | 4 | 296 | ||
AF099137.1 | - | 4 | 498 | |||
AF209747.1 | - | 4 | 705 | |||
AK298291.1 | - | 4 | 674 | |||
BC017825.1 | - | 4 | 696 | 4657825 |