Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019432.2 | NP_062305.1 | 2 | 666 | ||
GenBank mRNA | AJ272046.1 | - | 2 | 618 | ||
AK002424.1 | - | 2 | 685 | |||
AK050134.1 | - | 2 | 666 | |||
AK086093.1 | - | 2 | 671 | |||
BC024613.1 | - | 2 | 662 | 4215775 |