Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ywhag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018871.3 | NP_061359.2 | 2 | 1904 | ||
GenBank mRNA | AK088847.1 | - | 2 | 1818 | ||
AK153307.1 | - | 2 | 1861 | |||
AK164356.1 | - | 2 | 1855 | |||
AK179188.1 | - | 1 | 113 | |||
AK181145.1 | - | 1 | 113 | |||
AK191464.1 | - | 1 | 113 | |||
BC008129.1 | - | 2 | 1787 | 3498633 |