Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019493.3 | NP_062366.2 | 2, 3 | 395 | ||
GenBank mRNA | AB050983.2 | - | 2, 3 | 395 | ||
AJ005120.1 | - | 2, 3 | 268 | |||
AK136041.1 | - | 2, 3 | 382 | |||
AK136124.1 | - | 2, 3 | 383 | |||
AK136149.1 | - | 2, 3 | 385 | |||
AK136185.1 | - | 2, 3 | 387 | |||
AK161379.1 | - | 4, 5 | 474 | |||
AK162122.1 | - | 2, 3 | 382 | |||
AK163407.1 | - | 2, 3 | 381 | |||
BC066811.1 | - | 2, 3 | 369 | 30475849 |