Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fscn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019569.2 | NP_062515.2 | 1 | 196 | ||
GenBank mRNA | AF176024.2 | - | 1 | 308 | ||
BC050759.1 | - | 1 | 196 | 6770861 |