Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hebp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019487.3 | NP_062360.1 | 4 | 785 | ||
GenBank mRNA | AF117614.1 | - | 4 | 715 | ||
AK078130.1 | - | 4 | 785 | |||
BC006898.1 | - | 4 | 478 | 3599858 |