Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btg4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019493.3 | NP_062366.2 | 4, 5 | 740 | ||
GenBank mRNA | AB050983.2 | - | 4, 5 | 740 | ||
AJ005120.1 | - | 4, 5 | 613 | |||
AK135849.1 | - | 1, 2 | 119 | |||
AK136041.1 | - | 4, 5 | 727 | |||
AK136124.1 | - | 4, 5 | 728 | |||
AK136149.1 | - | 4, 5 | 730 | |||
AK136185.1 | - | 4, 5 | 732 | |||
AK161379.1 | - | 6, 7 | 819 | |||
AK163407.1 | - | 4, 5 | 728 | |||
BC066811.1 | - | 4, 5 | 714 | 30475849 |