Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Metap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019648.3 | NP_062622.1 | 2 | 796 | ||
GenBank mRNA | AB003144.1 | - | 1 | 107 | ||
AF434712.1 | - | 1 | 186 | |||
AK076804.1 | - | 1 | 139 | |||
AK090323.1 | - | 1 | 127 | |||
AK147097.1 | - | 1 | 201 | |||
AK159650.1 | - | 1 | 211 | |||
AK163318.1 | - | 1 | 213 | |||
AK165301.1 | - | 2 | 797 | |||
AK165845.1 | - | 1 | 191 | |||
BC002213.1 | - | 1 | 208 | 3490341 | ||
BC010263.1 | - | 1 | 181 | |||
BC092052.1 | - | 1 | 173 | 3670685 |