Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320350.1 | NP_001307279.1 | 30 | 4352 | ||
NM_001320351.1 | NP_001307280.1 | 28 | 4100 | |||
NM_001320352.1 | NP_001307281.1 | 4 | 464 | |||
NM_003307.3 | NP_003298.1 | 29 | 4202 | |||
NR_038257.1 | 29 | 4205 | ||||
XM_005261171.3 | XP_005261228.1 | 30 | 4363 | |||
XM_011529736.2 | XP_011528038.1 | 30 | 4696 | |||
XM_017028456.1 | XP_016883945.1 | 31 | 4456 | |||
XM_017028457.1 | XP_016883946.1 | 29 | 4595 | |||
GenBank mRNA | AB001535.1 | - | 29 | 4547 | ||
AB017549.1 | - | 5 | 433 | |||
AB166745.1 | - | 30 | 4315 | |||
AB166746.1 | - | 25 | 3554 | |||
AJ417076.1 | - | 29 | 3953 | |||
AJ878416.1 | - | 29 | 4165 | |||
AK301101.1 | - | 4 | 316 | |||
AK301451.1 | - | 10 | 1155 | |||
BC112342.1 | - | 30 | 4265 | 40069893 | ||
EU362987.1 | - | 4 | 464 |