Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh18a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019698.2 | NP_062672.2 | 5 | 686 | ||
NM_153554.2 | NP_705782.2 | 5 | 686 | |||
GenBank mRNA | AF056573.1 | - | 5 | 591 | ||
AF056574.1 | - | 5 | 591 | |||
AK134148.1 | - | 5 | 646 | |||
AK147711.1 | - | 5 | 606 | |||
AK151072.1 | - | 5 | 608 | |||
AK153946.1 | - | 5 | 607 | |||
AK159610.1 | - | 5 | 608 | |||
AK166459.1 | - | 5 | 571 | |||
AK166494.1 | - | 5 | 572 | |||
AK168905.1 | - | 5 | 606 | |||
AK211174.1 | - | 1 | 27 | |||
BC033427.1 | - | 5 | 617 | 5009115 | ||
BC037699.1 | - | 5 | 565 | 4188900 |