Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmnl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077698.1 | NP_001071166.1 | 3 | 480 | ||
NM_019679.2 | NP_062653.2 | 3 | 480 | |||
GenBank mRNA | AF006466.1 | - | 3 | 227 | ||
AF215666.1 | - | 3 | 305 |