Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Asah1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019734.2 | NP_062708.1 | 6, 7 | 575 | ||
GenBank mRNA | AF157500.1 | - | 6, 7 | 505 | ||
AF352179.1 | - | 6, 7 | 517 | |||
AK075666.1 | - | 6, 7 | 490 | |||
AK075919.1 | - | 6, 7 | 529 | |||
AK077721.1 | - | 6, 7 | 528 | |||
AK090314.1 | - | 6, 7 | 525 | |||
AK150567.1 | - | 6, 7 | 488 | |||
AK151208.1 | - | 6, 7 | 576 | |||
AK151312.1 | - | 6, 7 | 488 | |||
AK151478.1 | - | 6, 7 | 566 | |||
AK152305.1 | - | 6, 7 | 527 | |||
AK152811.1 | - | 6, 7 | 488 | |||
AK152920.1 | - | 6, 7 | 535 | |||
AK153101.1 | - | 6, 7 | 535 | |||
AK153297.1 | - | 6, 7 | 489 | |||
AK159556.1 | - | 6, 7 | 497 | |||
AK166757.1 | - | 6, 7 | 507 | |||
BC003204.1 | - | 6, 7 | 495 | 3587042 |